Learn More
Lead(II) chromate, 98%, Thermo Scientific Chemicals
$175.12 - $522.63
Chemical Identifiers
CAS | 7758-97-6 |
---|---|
Molecular Formula | CrO4Pb |
Molecular Weight (g/mol) | 323.192 |
MDL Number | MFCD00011158 |
InChI Key | MOUPNEIJQCETIW-UHFFFAOYSA-N |
Synonym | lead chromate, lead ii chromate, phoenicochroite, plumbous chromate, chrome yellow, chromate de plomb, lead chromate vi, lead chromate pbcro4, unii-aa3229aous |
PubChem CID | 24460 |
ChEBI | CHEBI:86257 |
IUPAC Name | dioxido(dioxo)chromium;lead(2+) |
SMILES | [O-][Cr](=O)(=O)[O-].[Pb+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1067830
|
Thermo Scientific Chemicals
A1067830 |
250 g |
Each for $175.12
|
|
||||||
AAA10678A1
|
Thermo Scientific Chemicals
A10678A1 |
1 kg |
Each for $522.63
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-97-6 | |
323.192 | |
MOUPNEIJQCETIW-UHFFFAOYSA-N | |
24460 | |
dioxido(dioxo)chromium;lead(2+) |
CrO4Pb | |
MFCD00011158 | |
lead chromate, lead ii chromate, phoenicochroite, plumbous chromate, chrome yellow, chromate de plomb, lead chromate vi, lead chromate pbcro4, unii-aa3229aous | |
CHEBI:86257 | |
[O-][Cr](=O)(=O)[O-].[Pb+2] |
Specifications
844°C | |
250 g | |
PbCrO4 | |
UN3077 | |
lead chromate, lead ii chromate, phoenicochroite, plumbous chromate, chrome yellow, chromate de plomb, lead chromate vi, lead chromate pbcro4, unii-aa3229aous | |
[O-][Cr](=O)(=O)[O-].[Pb+2] | |
323.192 | |
CHEBI:86257 | |
98% | |
6.3 |
7758-97-6 | |
CrO4Pb | |
MFCD00011158 | |
14,5405 | |
MOUPNEIJQCETIW-UHFFFAOYSA-N | |
dioxido(dioxo)chromium;lead(2+) | |
24460 | |
323.19 | |
Odorless | |
Lead (II) chromate |
Safety and Handling
GHS H Statement
H350-H360-H373-H201
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Explosive
mass explosion hazard.
P201-P202-P260-P281-P308+P313-P501c
H350-H360Df-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 231-846-0
RTECSNumber : GB2975000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only