missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lecithin, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210214791
Description
Molecular biology, Cell biologySpecifications
Lecithin | |
Yellow | |
1 kg | |
327.27 |
8030-76-0 | |
C11H22NO8P | |
L-a-Phosphatidylcholine | |
Solid |
Chemical Identifiers
8030-76-0 | |
C11H22NO8P | |
L-a-Phosphatidylcholine |
Safety and Handling
Recommended Storage : Store at 0°C.