missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lecithin, MP Biomedicals™
$88.40 - $582.69
Chemical Identifiers
CAS | 8030-76-0 |
---|---|
Molecular Formula | C11H22NO8P |
Synonym | L-a-Phosphatidylcholine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10214780
|
MP Biomedicals Inc
0210214780 |
100 g |
Each for $88.40
|
|
|||||
ICN10214791
|
MP Biomedicals Inc
0210214791 |
1 kg |
Each for $582.69
|
|
|||||
Description
Molecular biology, Cell biologyChemical Identifiers
8030-76-0 | |
C11H22NO8P | |
L-a-Phosphatidylcholine |
Specifications
8030-76-0 | |
C11H22NO8P | |
L-a-Phosphatidylcholine | |
Solid |
Yellow | |
100 g | |
327.27 | |
Lecithin |
Safety and Handling
Recommended Storage : Store at 0°C.