DWK Life Sciences Kimble™ Limited Volume Vial Insert
Inserts are ideal for limited and micro volume applications. Options are available for both standard and wide opening 12x32 autosampler type vials.
$68.32 - $550.49
Specifications
For Use With (Application) | limited and micro volume applications |
---|---|
Quantity | 100 |
Catalog Number | Mfr. No. | Volume (Metric) | Style | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Style | Price | Quantity | ||||
11-417-235
|
DWK Life Sciences
60850528 |
150 μL | Conical Point Design |
Case of 100 Each for $328.03
|
|
||||
11-417-236
|
DWK Life Sciences
60850629 |
300 μL | Conical Point Design |
Case of 100 Each for $550.49
|
|
||||
11-417-237
|
DWK Life Sciences
60855531 |
200 μL | Flat Bottom |
Case of 100 Each for $68.32
|
|
||||
11-417-238
|
DWK Life Sciences
60855631 |
400 μL | Flat Bottom |
Case of 100 Each for $72.82
|
|
||||
Description
- Clear borosilicate glass
- 60850 series has a conical point design, with polymer springs attached - residual volume 1 mL
- 60855 series has a flat bottom design - residual volume 25 μL
Specifications
limited and micro volume applications | |
100 |