Learn More
(+)-Limonene, 96%, unstabilized
CAS: 5989-27-5 | C10H16 | 136.24 g/mol
$94.40 - $566.98
Chemical Identifiers
| CAS | 5989-27-5 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00062991 |
| InChI Key | XMGQYMWWDOXHJM-JTQLQIEISA-N |
| Synonym | d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene |
| PubChem CID | 440917 |
| ChEBI | CHEBI:15382 |
| SMILES | CC(=C)[C@@H]1CCC(C)=CC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC179390050
|
Thermo Scientific Chemicals
179390050 |
5 mL | Glass bottle |
Each for $94.40
|
|
||||
|
AC179395000
|
Thermo Scientific Chemicals
179395000 |
500 mL | Glass Bottle |
Each for $146.43
|
|
||||
|
AC179390025
|
Thermo Scientific Chemicals
179390025 |
2.5 L | Glass bottle |
Each for $566.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5989-27-5 | |
| 136.24 | |
| XMGQYMWWDOXHJM-JTQLQIEISA-N | |
| 440917 | |
| CC(=C)[C@@H]1CCC(C)=CC1 |
| C10H16 | |
| MFCD00062991 | |
| d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene | |
| CHEBI:15382 |
Specifications
| 5989-27-5,10191-41-0 | |
| 100.0 | |
| Colorless to Yellow | |
| 175.4°C to 176.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4700 to 1.4740 | |
| 5 mL | |
| 0.8405 | |
| +106.00 (20.00°C c=1,CH3OH) | |
| d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene | |
| XMGQYMWWDOXHJM-JTQLQIEISA-N | |
| (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene | |
| 440917 | |
| CHEBI:15382 | |
| 96% | |
| (+)-Limonene |
| 95.0 | |
| -74.0°C | |
| 0.8405g/mL | |
| 48°C | |
| 95% min. (GC) | |
| C10H16 | |
| MFCD00062991 | |
| 05, 133 | |
| 14, 5493 | |
| + 106.00 | |
| Solubility in water: insoluble. Other solubilities: miscible with alcohol | |
| CC(=C)[C@@H]1CCC(C)=CC1 | |
| 136.24 | |
| 0.846 mPa.s (25°C) | |
| 136.24 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 227-813-5
RTECSNumber : GW6360000
TSCA : TSCA
RUO – Research Use Only