Learn More
(-)-Limonene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203735000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(-)-Limonene | |
95.0 | |
-104°C to 84°C | |
0.8420g/mL | |
48°C | |
95% min. (GC) | |
C10H16 | |
MFCD00001558 | |
05, 136 | |
14, 5493 | |
− 99.50 | |
Solubility in water: practically insoluble. Other solubilities: miscible with alcohol | |
CC1=CCC(CC1)C(=C)C | |
136.24 | |
CHEBI:15383 | |
96% |
5989-54-8 | |
100.0 | |
Colorless to Yellow | |
175.0°C to 177.0°C | |
Authentic | |
Glass bottle | |
1.4700 to 1.4740 | |
500 mL | |
0.842 | |
- 99.50 (20.00°C c=neat) | |
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s-- | |
XMGQYMWWDOXHJM-SNVBAGLBSA-N | |
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene | |
439250 | |
136.24 | |
Liquid |
Chemical Identifiers
5989-54-8 | |
136.24 | |
XMGQYMWWDOXHJM-SNVBAGLBSA-N | |
439250 | |
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene |
C10H16 | |
MFCD00001558 | |
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s-- | |
CHEBI:15383 | |
CC1=CCC(CC1)C(=C)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
GHS Signal Word: Warning
EINECSNumber : 227-815-6
RUO – Research Use Only