missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Limonene, 96%, Thermo Scientific Chemicals

$40.57 - $94.26

Chemical Identifiers

CAS 5989-54-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001558
InChI Key XMGQYMWWDOXHJM-SNVBAGLBSA-N
Synonym --limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
PubChem CID 439250
ChEBI CHEBI:15383
IUPAC Name (4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
SMILES CC1=CCC(CC1)C(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC203731000
SDS View Documents
Thermo Scientific Chemicals
203731000
100 mL Glass bottle
Each for $40.57
 
AC203735000
SDS View Documents
Thermo Scientific Chemicals
203735000
500 mL Glass bottle
Each for $94.26
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5989-54-8
136.24
XMGQYMWWDOXHJM-SNVBAGLBSA-N
439250
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
C10H16
MFCD00001558
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
CHEBI:15383
CC1=CCC(CC1)C(=C)C

Specifications

5989-54-8
100.0
Colorless to Yellow
175.0°C to 177.0°C
Authentic
Glass bottle
1.4700 to 1.4740
100 mL
0.842
- 99.50 (20.00°C c=neat)
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
XMGQYMWWDOXHJM-SNVBAGLBSA-N
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
439250
136.24
Liquid
95.0
-104°C to 84°C
0.8420g/mL
48°C
95% min. (GC)
C10H16
MFCD00001558
05, 136
14, 5493
− 99.50
Solubility in water: practically insoluble. Other solubilities: miscible with alcohol
CC1=CCC(CC1)C(=C)C
136.24
CHEBI:15383
96%
(-)-Limonene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.

GHS Signal Word: Warning

EINECSNumber : 227-815-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only