missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium, 99+%, granular, dry, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 211442500

 View more versions of this product

Catalog No. AC211442500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium
180.0°C
1340.0°C
99+%
MFCD00134051
01,570; 03,174; 04,286; 05,376; 10,234; 11,286; 13,157; 15,184; 17,161
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
WHXSMMKQMYFTQS-UHFFFAOYSA-N
lithium
3028194
6.94
Granular
99+%
7439-93-2
0.5300g/mL
Li
250 g
15, 5576
Solubility in water: reacts. Other solubilities: forming lioh and hydrogen, soluble in liquid ammonia and acids, decomposes in alcohol
[Li]
6.94
CHEBI:30145
≥99%

Chemical Identifiers

7439-93-2
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
lithium
Li
MFCD00134051
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
CHEBI:30145
[Li]
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-102-5

RTECSNumber : OJ5540000

TSCA : TSCA

Recommended Storage : product turns dark grey in air

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only