missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium, 99+%, granular, dry, Thermo Scientific Chemicals

$126.98 - $930.76

Chemical Identifiers

CAS 7439-93-2
Molecular Formula Li
Molecular Weight (g/mol) 6.94
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
PubChem CID 3028194
ChEBI CHEBI:30145
IUPAC Name lithium
SMILES [Li]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC211440250
SDS View Documents
Thermo Scientific Chemicals
211440250
25 g
Each for $126.98
 
AC211442500
SDS View Documents
Thermo Scientific Chemicals
211442500
250 g
Each for $930.76
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-93-2
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
lithium
Li
MFCD00134051
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
CHEBI:30145
[Li]

Specifications

180.0°C
1340.0°C
99+%
MFCD00134051
01,570; 03,174; 04,286; 05,376; 10,234; 11,286; 13,157; 15,184; 17,161
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
WHXSMMKQMYFTQS-UHFFFAOYSA-N
lithium
3028194
6.94
Granular
7439-93-2
0.5300g/mL
Li
25 g
15, 5576
Solubility in water: reacts. Other solubilities: forming lioh and hydrogen, soluble in liquid ammonia and acids, decomposes in alcohol
[Li]
6.94
CHEBI:30145
≥99%
Lithium, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-102-5

RTECSNumber : OJ5540000

TSCA : TSCA

Recommended Storage : product turns dark grey in air

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only