missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium acetate, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 297110250

 View more versions of this product

Catalog No. AC297110250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Specifications

Lithium acetate
546-89-4
25 g
C2H3LiO2
MFCD00013057
15,5577
Solubility in water: 408g/L (20°C). Other solubilities: 667g/L (40°C), 938g/L (50°C), freely soluble in alcohol
[Li+].CC([O-])=O
65.98
CHEBI:63045
≥99%
Authentic
280.0°C to 285.0°C
Solid
99+%
CH3COOLi
02,107
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
XIXADJRWDQXREU-UHFFFAOYSA-M
lithium;acetate
3474584
65.99
Analytical
Glass bottle

Chemical Identifiers

546-89-4
65.98
XIXADJRWDQXREU-UHFFFAOYSA-M
3474584
lithium;acetate
C2H3LiO2
MFCD00013057
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
CHEBI:63045
[Li+].CC([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-914-3

RTECSNumber : AI5450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only