Learn More
Lithium acetate, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
$73.64 - $185.69
Chemical Identifiers
CAS | 546-89-4 |
---|---|
Molecular Formula | C2H3LiO2 |
Molecular Weight (g/mol) | 65.98 |
MDL Number | MFCD00013057 |
InChI Key | XIXADJRWDQXREU-UHFFFAOYSA-M |
Synonym | lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt |
PubChem CID | 3474584 |
ChEBI | CHEBI:63045 |
IUPAC Name | lithium;acetate |
SMILES | [Li+].CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC297110250
|
Thermo Scientific Chemicals
297110250 |
25 g | Glass bottle |
Each for $73.64
|
|
||||
AC297111000
|
Thermo Scientific Chemicals
297111000 |
100 g | Glass bottle |
Each for $185.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
546-89-4 | |
65.98 | |
XIXADJRWDQXREU-UHFFFAOYSA-M | |
3474584 | |
lithium;acetate |
C2H3LiO2 | |
MFCD00013057 | |
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt | |
CHEBI:63045 | |
[Li+].CC([O-])=O |
Specifications
280.0°C to 285.0°C | |
Solid | |
99+% | |
CH3COOLi | |
02,107 | |
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt | |
XIXADJRWDQXREU-UHFFFAOYSA-M | |
lithium;acetate | |
3474584 | |
65.99 | |
Analytical | |
Glass bottle |
546-89-4 | |
25 g | |
C2H3LiO2 | |
MFCD00013057 | |
15,5577 | |
Solubility in water: 408g/L (20°C). Other solubilities: 667g/L (40°C), 938g/L (50°C), freely soluble in alcohol | |
[Li+].CC([O-])=O | |
65.98 | |
CHEBI:63045 | |
≥99% | |
Authentic | |
Lithium acetate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-914-3
RTECSNumber : AI5450000
TSCA : TSCA
RUO – Research Use Only