missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium acetylide ethylenediamine complex, 85%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 181251000

 View more versions of this product

Catalog No. AC181251000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Lithium acetylide ethylenediamine complex
6867-30-7
Yellow
80% min. (Gasvolumetric)
HC≡CLi·H2NCH2CH2NH2
100 g
lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
QJQWXTYPTBEPGS-UHFFFAOYSA-N
lithium;ethane-1,2-diamine;ethyne
2724010
85%
85%
76.0°C
110.6°C
C4H9LiN2
MFCD00013183
01,574; 05,382; 07,195; 11,288; 01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59
Solubility in water: vigorous reaction. Other solubilities: soluble in ethylene diamine,n-propylamine and,pyridine
[Li]C#C.NCCN
92.07
92.07
Crystalline Powder or Crystals

Chemical Identifiers

6867-30-7
92.07
QJQWXTYPTBEPGS-UHFFFAOYSA-N
2724010
[Li]C#C.NCCN
C4H9LiN2
MFCD00013183
lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
lithium;ethane-1,2-diamine;ethyne
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF SWAL

GHS Signal Word: Danger

EINECSNumber : 229-967-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only