Learn More
Lithium acetylide ethylenediamine complex, 85%, Thermo Scientific Chemicals
$463.77 - $463.77
Chemical Identifiers
CAS | 6867-30-7 |
---|---|
Molecular Formula | C4H9LiN2 |
Molecular Weight (g/mol) | 92.07 |
MDL Number | MFCD00013183 |
InChI Key | QJQWXTYPTBEPGS-UHFFFAOYSA-N |
Synonym | lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine |
PubChem CID | 2724010 |
IUPAC Name | lithium;ethane-1,2-diamine;ethyne |
SMILES | [Li]C#C.NCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC181251000
|
Thermo Scientific Chemicals
181251000 |
100 g |
Each for $463.77
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
6867-30-7 | |
92.07 | |
QJQWXTYPTBEPGS-UHFFFAOYSA-N | |
2724010 | |
[Li]C#C.NCCN |
C4H9LiN2 | |
MFCD00013183 | |
lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine | |
lithium;ethane-1,2-diamine;ethyne |
Specifications
6867-30-7 | |
Yellow | |
80% min. (Gasvolumetric) | |
HC≡CLi·H2NCH2CH2NH2 | |
100 g | |
lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine | |
QJQWXTYPTBEPGS-UHFFFAOYSA-N | |
lithium;ethane-1,2-diamine;ethyne | |
2724010 | |
85% | |
Lithium acetylide ethylenediamine complex, 85% |
76.0°C | |
110.6°C | |
C4H9LiN2 | |
MFCD00013183 | |
01,574; 05,382; 07,195; 11,288; 01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59 | |
Solubility in water: vigorous reaction. Other solubilities: soluble in ethylene diamine,n-propylamine and,pyridine | |
[Li]C#C.NCCN | |
92.07 | |
92.07 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF SWAL
GHS Signal Word: Danger
EINECSNumber : 229-967-9
TSCA : TSCA
RUO – Research Use Only