missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium benzoate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A165710I

 View more versions of this product

Catalog No. AAA165710I


Explore available promotions

Description

Description

In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

Solubility
Solubility in water is increased by sodium benzoate

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Lithium benzoate
>300°C
C7H5LiO2
5000 g
14,5523
Solubility in water is increased by sodium benzoate
[Li+].[O-]C(=O)C1=CC=CC=C1
128.06
128.06
553-54-8
Odorless
MFCD00035540
3598089
lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n
LDJNSLOKTFFLSL-UHFFFAOYSA-M
lithium(1+) benzoate
2724073
≥99%

Chemical Identifiers

553-54-8
128.06
LDJNSLOKTFFLSL-UHFFFAOYSA-M
2724073
C7H5LiO2
MFCD00035540
lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n
[Li+].[O-]C(=O)C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 209-042-6

RTECSNumber : OJ5720000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only