Learn More
Lithium benzoate, 99+%, Thermo Scientific Chemicals
$367.14 - $1173.93
Chemical Identifiers
CAS | 553-54-8 |
---|---|
Molecular Formula | C7H5LiO2 |
Molecular Weight (g/mol) | 128.06 |
MDL Number | MFCD00035540 |
InChI Key | LDJNSLOKTFFLSL-UHFFFAOYSA-M |
Synonym | lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n |
PubChem CID | 2724073 |
SMILES | [Li+].[O-]C(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1657130
|
Thermo Scientific Chemicals
A1657130 |
250 g |
Each for $367.14
|
|
|||||
AAA165710B
|
Thermo Scientific Chemicals
A165710B |
1000 g |
Each for $1,057.51
|
|
|||||
AAA165710I
|
Thermo Scientific Chemicals
A165710I |
5000 g |
Each for $1,173.93
|
|
|||||
Description
In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.
Solubility
Solubility in water is increased by sodium benzoate
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
553-54-8 | |
128.06 | |
LDJNSLOKTFFLSL-UHFFFAOYSA-M | |
2724073 |
C7H5LiO2 | |
MFCD00035540 | |
lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n | |
[Li+].[O-]C(=O)C1=CC=CC=C1 |
Specifications
553-54-8 | |
Odorless | |
MFCD00035540 | |
3598089 | |
lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n | |
LDJNSLOKTFFLSL-UHFFFAOYSA-M | |
lithium;benzoate | |
2724073 | |
≥99% |
>300°C | |
C7H5LiO2 | |
250 g | |
14,5523 | |
Solubility in water is increased by sodium benzoate | |
[Li+].[O-]C(=O)C1=CC=CC=C1 | |
128.06 | |
128.06 | |
Lithium benzoate |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 209-042-6
RTECSNumber : OJ5720000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only