missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01283918

 View more versions of this product

Catalog No. AA1283918

Explore available promotions

Description

Description

It act as a superconductor and widely used in lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium sulfide
12136-58-2
Powder
99.9% (Metal basis)
MFCD00011085
soluble in water and ethanol
[Li+].[Li+].[S--]
45.94
45.95
Moisture sensitive
1.66 g/cm3 at 20°C
900°C to 975°C
Yellow
50 g
Li2S
lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis
GLNWILHOFOBOFD-UHFFFAOYSA-N
dilithium(1+) sulfanediide
64734
Like rotten eggs
Yellow powder

Chemical Identifiers

12136-58-2
45.94
GLNWILHOFOBOFD-UHFFFAOYSA-N
64734
Li2S
MFCD00011085
lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis
[Li+].[Li+].[S--]
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H335

DOTInformation : DOT Class: 8; Packaging Group: II

EINECSNumber : 235-228-1

RTECSNumber : OJ6439500

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only