missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 358711000

 View more versions of this product

Catalog No. AC358711000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Lithium trifluoromethanesulfonate
33454-82-9
White or Colorless
4.34 to 4.56% (Li)
CF3LiO3S
MFCD00013227
lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
MCVFFRWZNYZUIJ-UHFFFAOYSA-M
lithium;trifluoromethanesulfonate
3664839
98%
98%
423.0°C
Authentic
Glass bottle
F3CSO3Li
100 g
Solubility in water: soluble
[Li+].C(F)(F)(F)S(=O)(=O)[O-]
156.01
156.01
Crystalline Powder or Crystals

Chemical Identifiers

33454-82-9
156.01
MCVFFRWZNYZUIJ-UHFFFAOYSA-M
3664839
[Li+].C(F)(F)(F)S(=O)(=O)[O-]
CF3LiO3S
MFCD00013227
lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
lithium;trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 251-528-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only