Learn More
Lithium trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
$72.12 - $500.53
Chemical Identifiers
CAS | 33454-82-9 |
---|---|
Molecular Formula | CF3LiO3S |
Molecular Weight (g/mol) | 156.01 |
MDL Number | MFCD00013227 |
InChI Key | MCVFFRWZNYZUIJ-UHFFFAOYSA-M |
Synonym | lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt |
PubChem CID | 3664839 |
IUPAC Name | lithium;trifluoromethanesulfonate |
SMILES | [Li+].C(F)(F)(F)S(=O)(=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC358710050
|
Thermo Scientific Chemicals
358710050 |
5 g | Glass bottle |
N/A
|
|
||||
AC358711000
|
Thermo Scientific Chemicals
358711000 |
100 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
33454-82-9 | |
156.01 | |
MCVFFRWZNYZUIJ-UHFFFAOYSA-M | |
3664839 | |
[Li+].C(F)(F)(F)S(=O)(=O)[O-] |
CF3LiO3S | |
MFCD00013227 | |
lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt | |
lithium;trifluoromethanesulfonate |
Specifications
33454-82-9 | |
White or Colorless | |
4.34 to 4.56% (Li) | |
CF3LiO3S | |
MFCD00013227 | |
lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt | |
MCVFFRWZNYZUIJ-UHFFFAOYSA-M | |
lithium;trifluoromethanesulfonate | |
3664839 | |
98% | |
Lithium trifluoromethanesulfonate, 98% |
423.0°C | |
Authentic | |
Glass bottle | |
F3CSO3Li | |
5 g | |
Solubility in water: soluble | |
[Li+].C(F)(F)(F)S(=O)(=O)[O-] | |
156.01 | |
156.01 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
EINECSNumber : 251-528-5
RUO – Research Use Only