missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Low Form Cylindrical Dewar Flask with Aluminum Housing

Curved bottom helps to increase coolant capacity

Supplier:  Fisherbrand™ FB1019621

 View more versions of this product

Catalog No. FB1019621


Explore available promotions

Description

Description

  • Cylindrical
  • Low Form
  • Aluminum Housing
  • Borosilicate Glass with Silvered Double Walls
Specifications

Specifications

70 mm
Liquid Nitrogen
54 mm
Aluminum
2.76 in.
125 mm
Silver
150 mL
SDS
Documents

Documents

Promotions

Promotions