missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Low Form Cylindrical Dewar Flask with Aluminum Housing
Curved bottom helps to increase coolant capacity
Supplier: Fisherbrand™ FB101967
Description
- Cylindrical
- Low Form
- Aluminum Housing
- Borosilicate Glass with Silvered Double Walls
Specifications
70 mm | |
Liquid Nitrogen | |
255 mm | |
Aluminum |
2.76 in. | |
325 mm | |
Silver | |
850 mL |