missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Lower Hemispherical Cloth Heating Mantle

Fisherbrand™ Lower Hemispherical Cloth Heating Mantle provides a safe and uniform heat distribution for round-bottom flasks.

 View more versions of this product

Catalog No. 11-476-004


Explore available promotions

Requires : Temperature Controller

Description

Description

  • Fiberglass cloth out shell molded to fit round-bottom flasks
  • Max temperature up to 482°C (900°F)
  • Uniform heat distribution
  • Multi-stranded resistance wire heating element for increased durability and flexibility
  • Temperature controller required for use

Warranty

1-year

Specifications

Specifications

RoHs, REACH
Round Bottom Flasks, Temperature Controller
76 mm (3 in.) lead wire with connector. 1.2 m (4 ft.) power adaptor cord with plug
Fiberglass
180 W
482°C (Max.)
8.5 oz
250 mL
120 V
Lower Hemispherical
Round/Bowl
Heating Mantle
10 A
Round bottom flask heating
Videos
SDS
Documents

Documents

Promotions

Promotions