BUCHI Lyovapor™ L-200 Freeze Dryer
The compact Lyovapor™ L-200 Classic offers high-quality Freeze Drying with a high level of automation.
Supplier:  BUCHI 200IN010200102
 View more versions of this product
Catalog No. 05403001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • A 6 kg condenser capacity allows for maximum sample throughput
  • Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime
  • Fully Automated
  • Method creation and reporting
  • Real-time pressure and temperature recording
  • Stoppering available under inert atmospheric conditions
  • Small footprint
  • Touch-screen control panel
Certifications
CE Certified
Specifications
Specifications
6 kg (Condensor)
Stainless Steel
Freeze Drying
23 in.
208/230 V
163 lb.
-55°C to +25°C
CSA
Manifold 12-Position, Mobile Trolley
20 in.
50/60 Hz
74 kg