BUCHI Lyovapor™ L-200 Freeze Dryer
The compact Lyovapor™ L-200 Classic offers high-quality Freeze Drying with a high level of automation.
$31301.27 - $45326.21
Specifications
Voltage 208/230 V
View More Specs
Products 6
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
05403001
View Documents
BUCHI
200IN010200102
Manifold 12-Position, Mobile Trolley N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
05405430
View Documents
BUCHI
11L200IN0102000002
Manifold Rack 12 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
05405431
View Documents
BUCHI
11L200IN0102001002
Manifold Rack 12, Pfieffer Duo 6 Vacuum Pump N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
05405432
View Documents
BUCHI
11L200IN0102001102
Manifold Rack 12, Pfieffer Duo 6 Vacuum Pump, Mobile Trolley N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
05405433
View Documents
BUCHI
11L200IN0102003002
Manifold Rack 12, Edwards nXDS6iC Vacuum Pump N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
05405434
View Documents
BUCHI
11L200IN1412003002
4 Shelves, Edwards nXDS6iC Vacuum Pump N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • A 6 kg condenser capacity allows for maximum sample throughput
  • Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime
  • Fully Automated
  • Method creation and reporting
  • Real-time pressure and temperature recording
  • Stoppering available under inert atmospheric conditions
  • Small footprint
  • Touch-screen control panel
Certifications
CE Certified
Specifications
Specifications
208/230 V
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'