Learn More
m-Cresol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110580010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
m-Cresol | |
8.0°C to 10.0°C | |
1.0300g/mL | |
86°C | |
98.5% min. (GC) | |
C7H8O | |
CH3C6H4OH | |
1 kg | |
06,373 | |
15,2564 | |
Solubility in water: in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform | |
CC1=CC(=CC=C1)O | |
108.14 | |
CHEBI:17231 | |
99% |
108-39-4 | |
Colorless to Red | |
203.0°C | |
Authentic | |
Glass Bottle | |
1.5390 to 1.5420 | |
MFCD00002302 | |
2076 | |
1.03 | |
m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol | |
RLSSMJSEOOYNOY-UHFFFAOYSA-N | |
3-methylphenol | |
342 | |
108.14 | |
Liquid |
Chemical Identifiers
108-39-4 | |
108.14 | |
RLSSMJSEOOYNOY-UHFFFAOYSA-N | |
342 | |
3-methylphenol |
C7H8O | |
MFCD00002302 | |
m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol | |
CHEBI:17231 | |
CC1=CC(=CC=C1)O |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 203-577-9
Recommended Storage : May darken on storage
RUO – Research Use Only