missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Cresol, 99%, Thermo Scientific Chemicals

$106.56 - $1488.81

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110581000
SDS View Documents
Thermo Scientific Chemicals
110581000
100 g Glass bottle
Each for $106.56
 
AC110585000
SDS View Documents
Thermo Scientific Chemicals
110585000
500 g Glass bottle
Each for $196.98
 
AC110580010
SDS View Documents
Thermo Scientific Chemicals
110580010
1 kg Glass Bottle
Each for $283.36
 
AC110580025
SDS View Documents
Thermo Scientific Chemicals
110580025
2.5 kg Glass bottle
Each for $423.98
 
AC110580100
SDS View Documents
Thermo Scientific Chemicals
110580100
10 kg Metal Drum
Each for $1,488.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-39-4
108.14
RLSSMJSEOOYNOY-UHFFFAOYSA-N
342
3-methylphenol
C7H8O
MFCD00002302
m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
CHEBI:17231
CC1=CC(=CC=C1)O

Specifications

108-39-4
Colorless to Red
203.0°C
Authentic
Glass bottle
1.5390 to 1.5420
MFCD00002302
06,373
15,2564
Solubility in water: in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
CC1=CC(=CC=C1)O
108.14
CHEBI:17231
99%
m-Cresol
8.0°C to 10.0°C
1.0300g/mL
86°C
98.5% min. (GC)
C7H8O
CH3C6H4OH
100 g
1.03
m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
RLSSMJSEOOYNOY-UHFFFAOYSA-N
3-methylphenol
342
108.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 203-577-9

Recommended Storage : May darken on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only