missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Cresol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 110580100

 View more versions of this product

Catalog No. AC110580100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

m-Cresol
8.0°C to 10.0°C
1.0300g/mL
86°C
98.5% min. (GC)
C7H8O
CH3C6H4OH
10 kg
1.03
m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
RLSSMJSEOOYNOY-UHFFFAOYSA-N
3-methylphenol
342
108.14
Liquid
108-39-4
Colorless to Red
203.0°C
Authentic
Metal Drum
1.5390 to 1.5420
MFCD00002302
06,373
15,2564
Solubility in water: in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
CC1=CC(=CC=C1)O
108.14
CHEBI:17231
99%

Chemical Identifiers

108-39-4
108.14
RLSSMJSEOOYNOY-UHFFFAOYSA-N
342
3-methylphenol
C7H8O
MFCD00002302
m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
CHEBI:17231
CC1=CC(=CC=C1)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 203-577-9

Recommended Storage : May darken on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only