missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Toluic acid, 99%, Thermo Scientific Chemicals

$45.38 - $176.54

Chemical Identifiers

CAS 99-04-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002523
InChI Key GPSDUZXPYCFOSQ-UHFFFAOYSA-N
Synonym m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
PubChem CID 7418
ChEBI CHEBI:10589
IUPAC Name 3-methylbenzoic acid
SMILES CC1=CC=CC(=C1)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC139050050
SDS View Documents
Thermo Scientific Chemicals
139050050
5 g Glass Bottle
Each for $50.20
 
AC139051000
SDS View Documents
Thermo Scientific Chemicals
139051000
100 g Plastic bottle
Each for $45.38
 
AC139050010
SDS View Documents
Thermo Scientific Chemicals
139050010
1 kg Plastic bottle
Each for $176.54
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99-04-7
136.15
GPSDUZXPYCFOSQ-UHFFFAOYSA-N
7418
3-methylbenzoic acid
C8H8O2
MFCD00002523
m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
CHEBI:10589
CC1=CC=CC(=C1)C(O)=O

Specifications

99-04-7
100.0
Beige-Yellow to Yellow
263°C
Authentic
Glass Bottle
CH3C6H4CO2H
5 g
14, 9535
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol and ether
CC1=CC=CC(=C1)C(O)=O
136.15
CHEBI:10589
99%
m-Toluic acid, 99%
98.5
108°C to 112°C
1.05
150°C
99%
C8H8O2
MFCD00002523
09, 475
m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
GPSDUZXPYCFOSQ-UHFFFAOYSA-N
3-methylbenzoic acid
7418
136.15
Crystalline Solid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-723-9

RTECSNumber : XU1200000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only