missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Tolylacetylene, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 392660050

 View more versions of this product

Catalog No. AC392660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

m-Tolylacetylene
96.0
0.9000g/mL
25°C
96% min. (GC)
C9H8
CH3C6H4C∽CH
5g
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
CC1=CC=CC(=C1)C#C
116.16
116.16
766-82-5
100.0
170°C to 175°C
Authentic
Glass bottle
1.5380 to 1.54
MFCD01318174
0.9
RENYIDZOAFFNHC-UHFFFAOYSA-N
1-ethynyl-3-methylbenzene
136600
97%

Chemical Identifiers

766-82-5
116.16
RENYIDZOAFFNHC-UHFFFAOYSA-N
136600
CC1=CC=CC(=C1)C#C
C9H8
MFCD01318174
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
1-ethynyl-3-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only