Learn More
m-Tolylacetylene, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 766-82-5 |
---|---|
Molecular Formula | C9H8 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD01318174 |
InChI Key | RENYIDZOAFFNHC-UHFFFAOYSA-N |
Synonym | 3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene |
PubChem CID | 136600 |
IUPAC Name | 1-ethynyl-3-methylbenzene |
SMILES | CC1=CC=CC(=C1)C#C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC392660010
|
Thermo Scientific Chemicals
392660010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC392660050
|
Thermo Scientific Chemicals
392660050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
766-82-5 | |
116.16 | |
RENYIDZOAFFNHC-UHFFFAOYSA-N | |
136600 | |
CC1=CC=CC(=C1)C#C |
C9H8 | |
MFCD01318174 | |
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene | |
1-ethynyl-3-methylbenzene |
Specifications
766-82-5 | |
100.0 | |
170°C to 175°C | |
Authentic | |
Glass bottle | |
1.5380 to 1.54 | |
MFCD01318174 | |
0.9 | |
RENYIDZOAFFNHC-UHFFFAOYSA-N | |
1-ethynyl-3-methylbenzene | |
136600 | |
97% |
96.0 | |
0.9000g/mL | |
25°C | |
96% min. (GC) | |
C9H8 | |
CH3C6H4C∽CH | |
1g | |
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene | |
CC1=CC=CC(=C1)C#C | |
116.16 | |
116.16 | |
m-Tolylacetylene |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
RUO â Research Use Only