missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega MagnaBot™ Magnetic Separation Devices DFS Item

Designed for high-throughput DNA purification when used with MagneSil™ paramagnetic particles

$1897.87 - $1897.87

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Magnesil™ paramagnetic particles
For Use With (Application) High-throughput bioseparation using Magnetic particles
Quantity 1/Ea.
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
PR-V3031 DFS Item
View Documents Promotion Details
Promega
V3031
Deep Well MagnaBot 96
Each for $1,897.87
 
Description

Description

Compatible with any plate-processing laboratory robot with a plate gripper.

Specifications

Specifications

15°C to 30°C
Magnesil™ paramagnetic particles
High-throughput bioseparation using Magnetic particles
1/Ea.
SDS
missing translation for 'documents'

missing translation for 'documents'