Promega MagnaBot™ Magnetic Separation Devices
Designed for high-throughput DNA purification when used with MagneSil™ paramagnetic particles
Supplier: Promega V3031
Description
- Designed for high-throughput bioseparation in deep-well plates using magnetic particles
- An alternative to vacuum filtration and centrifugation separation formats
- Use for automated nucleic acid purification applications
Specifications
15°C to 30°C | |
High-throughput bioseparation using Magnetic particles | |
1/Ea. |
Magnesil™ paramagnetic particles | |
Deep Well MagnaBot 96 |