missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium bromide ethyl etherate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 209701000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium bromide ethyl etherate | |
Beige to White | |
Authentic | |
Glass bottle | |
(C2H5)2O·MgBr2 | |
100 g | |
magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent | |
JGZKUKYUQJUUNE-UHFFFAOYSA-L | |
magnesium;ethoxyethane;dibromide | |
11010650 | |
99% |
29858-07-9 | |
35°C | |
98.5% min. (Argentometry) | |
C4H10Br2MgO | |
MFCD00064500 | |
01,629; 07,218; 13,259; 17,174 | |
Solubility in water: soluble | |
Br[Mg]Br.CCOCC | |
258.24 | |
258.24 | |
Crystalline Powder and/or Chunks |
Chemical Identifiers
29858-07-9 | |
258.24 | |
JGZKUKYUQJUUNE-UHFFFAOYSA-L | |
11010650 | |
Br[Mg]Br.CCOCC |
C4H10Br2MgO | |
MFCD00064500 | |
magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent | |
magnesium;ethoxyethane;dibromide |
Safety and Handling
GHS H Statement
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
RUO – Research Use Only