missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium bromide ethyl etherate, 99%, Thermo Scientific Chemicals

$348.47 - $348.47

Chemical Identifiers

CAS 29858-07-9
Molecular Formula C4H10Br2MgO
Molecular Weight (g/mol) 258.24
MDL Number MFCD00064500
InChI Key JGZKUKYUQJUUNE-UHFFFAOYSA-L
Synonym magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
PubChem CID 11010650
IUPAC Name magnesium;ethoxyethane;dibromide
SMILES Br[Mg]Br.CCOCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC209701000
SDS View Documents
Thermo Scientific Chemicals
209701000
100 g Glass bottle
Each for $348.47
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29858-07-9
258.24
JGZKUKYUQJUUNE-UHFFFAOYSA-L
11010650
Br[Mg]Br.CCOCC
C4H10Br2MgO
MFCD00064500
magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
magnesium;ethoxyethane;dibromide

Specifications

29858-07-9
35°C
98.5% min. (Argentometry)
C4H10Br2MgO
MFCD00064500
01,629; 07,218; 13,259; 17,174
Solubility in water: soluble
Br[Mg]Br.CCOCC
258.24
258.24
Crystalline Powder and/or Chunks
Beige to White
Authentic
Glass bottle
(C2H5)2O·MgBr2
100 g
magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
JGZKUKYUQJUUNE-UHFFFAOYSA-L
magnesium;ethoxyethane;dibromide
11010650
99%
Magnesium bromide ethyl etherate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only