missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium Chloride Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific M333
Specifications
Magnesium Chloride Hexahydrate | |
White | |
5 to 6.5 | |
3 kg | |
Cl2H12MgO6 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
magnesium(2+) hexahydrate dichloride | |
203.3 | |
Certified ACS | |
Poly Pail | |
1.56g/mL; |
117°C | |
7791-18-6 | |
Powder/Solid | |
99 to 102 % | |
MFCD00149781 | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
203.30 | |
99 to 102% | |
Pass Test | |
>106°C |
Chemical Identifiers
7791-18-6 | |
203.30 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Cl2H12MgO6 | |
MFCD00149781 | |
magnesium(2+) hexahydrate dichloride |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and respiratory system. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1