missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™

$333.25 - $1151.08

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M33-500 Edge
SDS View Documents Promotion Details
Fisher Chemical
M33500
500 g Poly Bottle
Each for $333.25
Case of 6 Each for $1,999.50
Only null left
Edge
 
M33-3
SDS View Documents Promotion Details
Fisher Chemical
M333
3 kg Poly Pail
Each for $1,151.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7791-18-6
203.30
DHRRIBDTHFBPNG-UHFFFAOYSA-L
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
Cl2H12MgO6
MFCD00149781
magnesium(2+) hexahydrate dichloride

Specifications

117°C
7791-18-6
Powder/Solid
99 to 102%
MFCD00149781
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
203.30
99 to 102%
Pass Test
>106°C
Magnesium Chloride Hexahydrate
White
5 to 6.5
500 g
Cl2H12MgO6
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
203.3
Certified ACS
Poly Bottle
1.56g/mL
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and respiratory system. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1

SDS
missing translation for 'documents'

missing translation for 'documents'