Learn More
Magnesium chloride, pure, Thermo Scientific Chemicals
$69.83 - $975.17
Chemical Identifiers
CAS | 7786-30-3 |
---|---|
Molecular Formula | Cl2Mg |
Molecular Weight (g/mol) | 95.21 |
MDL Number | MFCD00011106 |
InChI Key | TWRXJAOTZQYOKJ-UHFFFAOYSA-L |
Synonym | magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride |
PubChem CID | 5360315 |
ChEBI | CHEBI:6636 |
SMILES | [Mg++].[Cl-].[Cl-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC223211000
|
Thermo Scientific Chemicals
223211000 |
100 g | Plastic bottle |
Each for $69.83
|
|
||||
AC223210010
|
Thermo Scientific Chemicals
223210010 |
1 kg | Plastic bottle |
Each for $106.36
|
|
||||
AC223210025
|
Thermo Scientific Chemicals
223210025 |
2.5 kg | Plastic bottle |
Each for $214.30
|
|
||||
AC223210250
|
Thermo Scientific Chemicals
223210250 |
25 kg | Fibre drum |
Each for $975.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7786-30-3 | |
95.21 | |
TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
5360315 | |
[Mg++].[Cl-].[Cl-] |
Cl2Mg | |
MFCD00011106 | |
magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
CHEBI:6636 |
Specifications
1412.0°C | |
7786-30-3 | |
Powder | |
Cl2Mg | |
MFCD00011106 | |
15,5726 | |
Solubility in water: 540g/L (20°C) | |
[Mg++].[Cl-].[Cl-] | |
95.21 | |
CHEBI:6636 | |
≥98.0% (on dry substance) | |
1.5% max. (105°C, 2 Hrs) | |
Magnesium chloride |
708.0°C | |
White | |
100 g | |
MgCl2 | |
11,31 | |
magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
magnesium;dichloride | |
5360315 | |
95.21 | |
Pure | |
Plastic bottle |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 232-094-6
RUO – Research Use Only