Learn More
Magnesium chromate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals
Corrosion inhibitor, pigment
Supplier: Thermo Scientific Chemicals 04050336
Description
Used as Corrosion inhibitor, pigment. As an ingredient in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium chromate hydrate | |
Crystalline | |
(metals basis) | |
MFCD00801009 | |
Magnesium chromate | |
CRGGPIWCSGOBDN-UHFFFAOYSA-N | |
magnesium(2+) dioxochromiumbis(olate) | |
140.30 (Anhydrous) |
16569-85-0 | |
500 g | |
CrMgO4 | |
UN2923 | |
Soluble in water | |
[Mg++].[O-][Cr]([O-])(=O)=O | |
140.30 | |
99.8% |
Chemical Identifiers
16569-85-0 | |
140.30 | |
CRGGPIWCSGOBDN-UHFFFAOYSA-N | |
magnesium(2+) dioxochromiumbis(olate) |
CrMgO4 | |
MFCD00801009 | |
Magnesium chromate | |
[Mg++].[O-][Cr]([O-])(=O)=O |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P362-P501c
H301-H312-H315-H317-H318-H335-H340-H350
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only