missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium chromate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals

Corrosion inhibitor, pigment

$332.37 - $332.37

Chemical Identifiers

CAS 16569-85-0
Molecular Formula CrMgO4
Molecular Weight (g/mol) 140.30
MDL Number MFCD00801009
InChI Key CRGGPIWCSGOBDN-UHFFFAOYSA-N
Synonym Magnesium chromate
IUPAC Name magnesium(2+) dioxochromiumbis(olate)
SMILES [Mg++].[O-][Cr]([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4050336
View Documents
Thermo Scientific Chemicals
04050336
500 g
Each for $332.37
 
Description

Description

Used as Corrosion inhibitor, pigment. As an ingredient in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16569-85-0
140.30
CRGGPIWCSGOBDN-UHFFFAOYSA-N
magnesium(2+) dioxochromiumbis(olate)
CrMgO4
MFCD00801009
Magnesium chromate
[Mg++].[O-][Cr]([O-])(=O)=O

Specifications

16569-85-0
500 g
MFCD00801009
Magnesium chromate
CRGGPIWCSGOBDN-UHFFFAOYSA-N
magnesium(2+) dioxochromiumbis(olate)
140.30 (Anhydrous)
Magnesium chromate hydrate
Crystalline
CrMgO4
UN2923
Soluble in water
[Mg++].[O-][Cr]([O-])(=O)=O
140.30
99.8%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P362-P501c

H301-H312-H315-H317-H318-H335-H340-H350

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only