missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041921A7

 View more versions of this product

Catalog No. AA41921A7


Explore available promotions

Description

Description

Used as a laxative or dietary supplement

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium citrate hydrate
3344-18-1
5 kg
MFCD09953657
trimagnesium 2+ dicitrate hydrate
HZTVQLXIRHEMMB-UHFFFAOYSA-H
trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
122198783
Mg ≈ 16% (dry wt.)
White
Powder
C12H12Mg3O15
14,5663
Slightly soluble in water.
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2]
469.128
451.13 (Anhydrous)

Chemical Identifiers

3344-18-1
469.128
HZTVQLXIRHEMMB-UHFFFAOYSA-H
122198783
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2]
C12H12Mg3O15
MFCD09953657
trimagnesium 2+ dicitrate hydrate
trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 222-093-9

RTECSNumber : TZ8552900

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only