missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals
$107.92 - $357.93
Chemical Identifiers
CAS | 3344-18-1 |
---|---|
Molecular Formula | C12H12Mg3O15 |
Molecular Weight (g/mol) | 469.128 |
MDL Number | MFCD09953657 |
InChI Key | HZTVQLXIRHEMMB-UHFFFAOYSA-H |
Synonym | trimagnesium 2+ dicitrate hydrate |
PubChem CID | 122198783 |
IUPAC Name | trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate |
SMILES | C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2] |
Description
Used as a laxative or dietary supplement
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3344-18-1 | |
469.128 | |
HZTVQLXIRHEMMB-UHFFFAOYSA-H | |
122198783 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2] |
C12H12Mg3O15 | |
MFCD09953657 | |
trimagnesium 2+ dicitrate hydrate | |
trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate |
Specifications
White | |
Powder | |
C12H12Mg3O15 | |
14,5663 | |
Slightly soluble in water. | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2] | |
469.128 | |
451.13 (Anhydrous) | |
Magnesium citrate hydrate |
3344-18-1 | |
1 kg | |
MFCD09953657 | |
trimagnesium 2+ dicitrate hydrate | |
HZTVQLXIRHEMMB-UHFFFAOYSA-H | |
trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate | |
122198783 | |
Mg ≈ 16% (dry wt.) |
Safety and Handling
EINECSNumber : 222-093-9
RTECSNumber : TZ8552900
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only