missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium phosphate, tribasic hydrate, 98+%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 391942500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium phosphate, tribasic hydrate | |
53408-95-0 | |
250 g | |
Mg3O8P2 | |
magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate | |
GVALZJMUIHGIMD-UHFFFAOYSA-H | |
trimagnesium(2+) diphosphate | |
6096954 | |
≥98% | |
32.0% max. (425°C) |
White | |
Powder | |
98+% | |
MFCD00150251 | |
Solubility in water: insoluble. | |
[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
262.86 | |
262.86 | |
Pure | |
Plastic bottle |
Chemical Identifiers
53408-95-0 | |
262.86 | |
GVALZJMUIHGIMD-UHFFFAOYSA-H | |
6096954 |
Mg3O8P2 | |
MFCD00150251 | |
magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate | |
[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
RUO – Research Use Only