missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium phosphate, tribasic hydrate, 98+%, pure, Thermo Scientific Chemicals

$54.04 - $54.04

Chemical Identifiers

CAS 53408-95-0
Molecular Formula Mg3O8P2
Molecular Weight (g/mol) 262.86
MDL Number MFCD00150251
InChI Key GVALZJMUIHGIMD-UHFFFAOYSA-H
Synonym magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
PubChem CID 6096954
SMILES [Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391942500
SDS View Documents
Thermo Scientific Chemicals
391942500
250 g Plastic bottle
Each for $54.04
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53408-95-0
262.86
GVALZJMUIHGIMD-UHFFFAOYSA-H
6096954
Mg3O8P2
MFCD00150251
magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

White
Powder
98+%
MFCD00150251
Solubility in water: insoluble.
[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
262.86
262.86
Pure
Plastic bottle
53408-95-0
250 g
Mg3O8P2
magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
GVALZJMUIHGIMD-UHFFFAOYSA-H
trimagnesium(2+) diphosphate
6096954
≥98%
32.0% max. (425°C)
Magnesium phosphate, tribasic hydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only