missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium stearate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A162440B

 View more versions of this product

Catalog No. AAA162440B


Explore available promotions

Description

Description

Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ing
Specifications

Specifications

Magnesium stearate
148°C to 152°C
C36H70MgO4
1000 g
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
HQKMJHAJHXVSDF-UHFFFAOYSA-L
magnesium;octadecanoate
11177
591.27
557-04-0
Odorless
MFCD00036391
14,5690
Slightly soluble in benzene. Insoluble in water,alcohol and ether.
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]
591.257
CHEBI:9254

Chemical Identifiers

557-04-0
591.257
HQKMJHAJHXVSDF-UHFFFAOYSA-L
11177
magnesium;octadecanoate
C36H70MgO4
MFCD00036391
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
CHEBI:9254
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-150-3

RTECSNumber : WI4390000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only