Learn More
Magnesium stearate, Thermo Scientific Chemicals
$96.08 - $291.60
Chemical Identifiers
CAS | 557-04-0 |
---|---|
Molecular Formula | C36H70MgO4 |
Molecular Weight (g/mol) | 591.257 |
MDL Number | MFCD00036391 |
InChI Key | HQKMJHAJHXVSDF-UHFFFAOYSA-L |
Synonym | magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt |
PubChem CID | 11177 |
ChEBI | CHEBI:9254 |
IUPAC Name | magnesium;octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1624430
|
Thermo Scientific Chemicals
A1624430 |
250 g |
Each for $96.08
|
|
|||||
AAA162440B
|
Thermo Scientific Chemicals
A162440B |
1000 g |
Each for $291.60
|
|
|||||
Description
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMagnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ing
Chemical Identifiers
557-04-0 | |
591.257 | |
HQKMJHAJHXVSDF-UHFFFAOYSA-L | |
11177 | |
magnesium;octadecanoate |
C36H70MgO4 | |
MFCD00036391 | |
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt | |
CHEBI:9254 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2] |
Specifications
557-04-0 | |
C36H70MgO4 | |
250 g | |
Slightly soluble in benzene. Insoluble in water,alcohol and ether. | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2] | |
591.257 | |
CHEBI:9254 | |
Magnesium stearate |
148°C to 152°C | |
MFCD00036391 | |
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt | |
HQKMJHAJHXVSDF-UHFFFAOYSA-L | |
magnesium;octadecanoate | |
11177 | |
591.27 |
Safety and Handling
EINECSNumber : 209-150-3
RTECSNumber : WI4390000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only