Idex Male-Threaded Nuts DFS Item
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Supplier:  Idex UPU410X
 View more versions of this product
Catalog No. 05-700-233
Explore available promotions
Description
Description
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Specifications
Specifications
0.31 in.
1/16 in. O.D. Tubing
Stainless Steel
0.46 in.
0.0625 in.
Nut