Idex Male-Threaded Nuts
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
$90.09 - $133.64
Catalog Number | Mfr. No. | For Use With (Equipment) | Length (English) | Material | Width (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Length (English) | Material | Width (English) | Price | Quantity | ||
05-700-233
|
Idex
UPU410X |
1/16 in. O.D. Tubing | 0.46 in. | Stainless Steel | 0.31 in. |
Pack of 10 Each for $133.64
|
|
||
05-700-229
|
Idex
UPU400X |
1/16 in. O.D. Tubing | 0.4 in. | Stainless Steel | 0.25 in. |
Pack of 10 Each for $124.47
|
|
||
05-700-96
|
Idex
UPP255X |
1/32 in. or 1/16 in. OD Tubing | 0.87 in. | PEEK | 0.365 in. |
Pack of 10 Each for $90.09
|
|
||
Description
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
- Eliminates tubing twist that can loosen connections
- Resistant to loosening through vibration