Learn More
Malonic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125262500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Malonic acid | |
130.0°C to 135.0°C | |
172°C | |
99% | |
C3H4O4 | |
250 g | |
15,5774 | |
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in alcohol and ether | |
C(C(=O)O)C(=O)O | |
104.06 | |
CHEBI:30794 | |
99% |
141-82-2 | |
White | |
Authentic | |
Plastic bottle | |
HO2CCH2CO2H | |
02,566 | |
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid | |
OFOBLEOULBTSOW-UHFFFAOYSA-N | |
propanedioic acid | |
867 | |
104.06 | |
Crystalline Powder |
Chemical Identifiers
141-82-2 | |
104.06 | |
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid | |
CHEBI:30794 | |
C(C(=O)O)C(=O)O |
C3H4O4 | |
OFOBLEOULBTSOW-UHFFFAOYSA-N | |
867 | |
propanedioic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 205-503-
RUO – Research Use Only