missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid, 99%

CAS: 141-82-2 | C3H4O4 | 104.06 g/mol

$226.55 - $590.79

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3H4O4
Molecular Weight (g/mol) 104.06
InChI Key OFOBLEOULBTSOW-UHFFFAOYSA-N
Synonym malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
PubChem CID 867
ChEBI CHEBI:30794
IUPAC Name propanedioic acid
SMILES C(C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125262500
SDS View Documents
Thermo Scientific Chemicals
125262500
250 g Plastic bottle
Each for $226.55
Only null left
 
AC125260010
SDS View Documents
Thermo Scientific Chemicals
125260010
1 kg Plastic bottle
Each for $590.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-82-2
104.06
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
CHEBI:30794
C(C(=O)O)C(=O)O
C3H4O4
OFOBLEOULBTSOW-UHFFFAOYSA-N
867
propanedioic acid

Specifications

141-82-2
White
Authentic
Plastic bottle
HO2CCH2CO2H
02,566
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
OFOBLEOULBTSOW-UHFFFAOYSA-N
propanedioic acid
867
104.06
Crystalline Powder
130.0°C to 135.0°C
172°C
99%
C3H4O4
250 g
15,5774
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in alcohol and ether
C(C(=O)O)C(=O)O
104.06
CHEBI:30794
99%
Malonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 205-503-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only