Learn More
Malonic acid dihydrazide, 99%, Thermo Scientific Chemicals
$96.27 - $378.42
Chemical Identifiers
CAS | 3815-86-9 |
---|---|
Molecular Formula | C3H8N4O2 |
Molecular Weight (g/mol) | 132.12 |
MDL Number | MFCD00041268 |
InChI Key | PSIKPHJLTVSQFO-UHFFFAOYSA-N |
Synonym | malonic acid dihydrazide, malonohydrazide, malonic dihydrazide, propanedioic acid, dihydrazide, malonhydrazide, malonyl dihydrazide, malonic acid, dihydrazide, malonyl hydrazide, malonic acid hydrazide, malonoyldihydrazine |
PubChem CID | 77433 |
IUPAC Name | propanedihydrazide |
SMILES | NNC(=O)CC(=O)NN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1227006
|
Thermo Scientific Chemicals
L1227006 |
5 g |
Each for $96.27
|
|
|||||
AAL1227014
|
Thermo Scientific Chemicals
L1227014 |
25 g |
Each for $378.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3815-86-9 | |
132.12 | |
PSIKPHJLTVSQFO-UHFFFAOYSA-N | |
77433 | |
NNC(=O)CC(=O)NN |
C3H8N4O2 | |
MFCD00041268 | |
malonic acid dihydrazide, malonohydrazide, malonic dihydrazide, propanedioic acid, dihydrazide, malonhydrazide, malonyl dihydrazide, malonic acid, dihydrazide, malonyl hydrazide, malonic acid hydrazide, malonoyldihydrazine | |
propanedihydrazide |
Specifications
3815-86-9 | |
C3H8N4O2 | |
5 g | |
malonic acid dihydrazide, malonohydrazide, malonic dihydrazide, propanedioic acid, dihydrazide, malonhydrazide, malonyl dihydrazide, malonic acid, dihydrazide, malonyl hydrazide, malonic acid hydrazide, malonoyldihydrazine | |
NNC(=O)CC(=O)NN | |
132.12 | |
132.12 | |
Malonic acid dihydrazide |
152°C to 156°C | |
MFCD00041268 | |
1072337 | |
PSIKPHJLTVSQFO-UHFFFAOYSA-N | |
propanedihydrazide | |
77433 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
RTECSNumber : OO8000000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only