Learn More
Malonyl dichloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125281000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Malonyl dichloride | |
Brown to Yellow | |
47.0°C (15.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4620 to 1.4660 | |
MFCD00000735 | |
02, I, 252 | |
1.45 | |
Solubility in water: reacts violently. Other solubilities: soluble in ether | |
C(C(=O)Cl)C(=O)Cl | |
140.95 | |
140.95 | |
Liquid |
1663-67-8 | |
1.4500g/mL | |
47°C | |
96% min. (Argentometry) | |
C3H2Cl2O2 | |
ClCOCH2COCl | |
100 g | |
12,291 | |
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride | |
SXYFKXOFMCIXQW-UHFFFAOYSA-N | |
propanedioyl dichloride | |
74269 | |
97% |
Chemical Identifiers
1663-67-8 | |
140.95 | |
SXYFKXOFMCIXQW-UHFFFAOYSA-N | |
74269 | |
C(C(=O)Cl)C(=O)Cl |
C3H2Cl2O2 | |
MFCD00000735 | |
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride | |
propanedioyl dichloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 216-772-9
RUO – Research Use Only